missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STXBP6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10976-4-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
STXBP6 binds components of the SNARE complex.Specifica
STXBP6 | |
Polyclonal | |
Unconjugated | |
STXBP6 | |
amisyn, HSPC156, STXBP6, Syntaxin binding protein 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217517, 29091, 362734 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8NFX7, Q8R3T5 | |
STXBP6 | |
STXBP6 Fusion Protein Ag1427 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |