missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STYK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18028-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
STYK1, a probable tyrosine protein-kinase, which has strong transforming capabilities on a variety of cell lines. When overexpressed, it can also induce tumor cell invasion as well as metastasis in distant organs. May act by activating both MAP kinase and phosphatidylinositol 3′-kinases (PI3K) pathways. It is widely expressed; highly expressed in brain, placenta and prostate. STYK1 is expressed in tumor cells such as hepatoma cells LO2, cervix carcinoma cells HeLa, ovary cancer cells Ho8910 and chronic myelogenous leukemia cells K562, but not in other tumor cells such as epidermoid carcinoma.Specifica
STYK1 | |
Polyclonal | |
Unconjugated | |
STYK1 | |
NOK, Protein PK unique, STYK1, SuRTK106, Tyrosine protein kinase STYK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
243659, 500340, 55359 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6J9G0, Q6J9G1 | |
STYK1 | |
STYK1 Fusion Protein Ag12594 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |