Learn More
Cytiva (formerly Pall Lab) Supor™ 100 Membrane Disc Filters
Membranes are used to sterilize liquid reagents, remove particulate contamination, and clarify solutions prior to further processing.
Marca: Cytiva (formerly Pall Lab) 60309
Dettagli aggiuntivi : Peso : 0.49400kg
Descrizione
- These products are optimized for biological, pharmaceutical and sterilizing filtration requirements
- For research use only
Specifica
PES | |
Non-sterile | |
Plain |
0.1 μm | |
132 μm | |
25 mm |