missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 10 Mouse anti-Human, Clone: 1B1C3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66821-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
SNARE involved in vesicular transport from the late endosomes to the trans-Golgi network.Specifica
Syntaxin 10 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60499 | |
STX10 | |
Syntaxin 10 Fusion Protein Ag27861 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
1B1C3 | |
Unconjugated | |
STX10 | |
hsyn10, STX10, SYN10, syntaxin 10 | |
Mouse | |
Protein G | |
RUO | |
8677 | |
-20°C | |
Liquid |