Learn More
BRAND™ Tamper-Evident Screw Cap
Tamper-Evident Screw Cap equipped with a silicone seal is suitable for use with microtubes.
Marca: BRAND™ 780775
147.62 EUR valido fino al 2024-12-31
Usa il codice promo "23264" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.44000kg
Descrizione
- With silicone seal
- For microtubes
- Working temperature between -196 to 121°C
Specifica
Silicone Seal | |
Silicone | |
Yellow |
Tamper Evident Screw Cap | |
Polypropylene | |
78076064 Microtubes |