Learn More
Brady™ Tamper-evident Thermal Transfer Vinyl Labels
Printable, tamper-evident vinyl labels for Thermal Transfer Printers.
Marca: Brady™ 030173
Dettagli aggiuntivi : Peso : 1.40000kg
Descrizione
- Designed to fracture easily in order to show signs of product tampering and prevent one-piece label removal
- Use caution when removing from liner, as material is fragile
Specifica
Not Autoclavable | |
White | |
Tamper-resistant Vinyl | |
Non-sterile | |
BBP™11, BradyPrinter i5100, i7100, BradyPrinter PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers | |
5 | |
Rectangle | |
0.066 mm | |
12.7 mm |
Permanent Acrylic | |
Satin | |
Thermal Transfer | |
Circuit Board Labeling, Security Identification | |
16.8 mm | |
10,000 Labels | |
-40°C to +80°C | |
13.6 mm | |
11.1 mm |