Learn More
TBS, Tris Buffered Saline, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents
Marca: Fisher Bioreagents BP2472-100
Dettagli aggiuntivi : CAS : 7732-18-5 Peso : 0.15000kg
Descrizione
Tris Buffered Saline (TBS) is used in biochemical and molecular biology applications.Specifica
Tris Buffered Saline | |
7647-14-5,7447-40-7,7647-01-0,7732-18-5,77-86-1 | |
Colorless | |
Liquid | |
DNase free | |
Protease free | |
Molecular Biology |
1X Solution, pH 7.4 | |
100°C | |
7.5 | |
100 mL | |
Poly Bottle | |
TBS |