missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEAL7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11218-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Plays a role in the negative regulation of NF-kappa-B signaling at the basal level by modulating transcriptional activity of NF-kappa-B on its target gene promoters. Associates with cyclin D1 promoter containing Myc E-box sequence and transcriptionally represses cyclin D1 expression. Regulates telomerase reverse transcriptase expression and telomerase activity in both ALT (alternative lengthening of telomeres)and telomerase-positive cell lines.Specifica
TCEAL7 | |
Polyclonal | |
Unconjugated | |
TCEAL7 | |
MPMGp800C04260Q003, TCEA like protein 7, TCEAL7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56849 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BRU2 | |
TCEAL7 | |
TCEAL7 Fusion Protein Ag1715 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |