missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12157-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Transcobalamin I (TCI) and Transcobalamin II (TCII) are secreted proteins belonging to the eukaryotic cobalamin transport proteins family and also to the vitamin B12-binding protein family. The genes encoding these proteins map to chromosome 11q11-q12 and 22q12.2, respectively. Transcobalamin I is a constituent of secondary granules in neutrophils, while Transcobalamin II binds cobalamin and mediates its transport into cells. These plasma proteins are expressed in various tissues and secretions.Specifica
TCN2 | |
Polyclonal | |
Unconjugated | |
TCN2 | |
D22S676, D22S750, TC 2, TC II, TC2, TCII, TCN2, Transcobalamin 2, Transcobalamin II | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21452, 64365, 6948 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88968, P20062, Q9R0D6 | |
Tcn2 | |
TCN2 Fusion Protein Ag2825 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |