missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR beta Armenian Hamster, PerCP-Cy5.5, Clone: H57-597, enQuireBio™
Armenian Hamster Monoclonal Antibody
Marca: EnquireBio QAB79-PCP55-25ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
TCR beta | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Armenian Hamster | |
25 μg | |
Primary | |
Store at 2-4°C. |
Flow Cytometry | |
H57-597 | |
PerCP-Cyanine5.5 | |
Tcrb | |
Protein A/G Purified | |
RUO | |
21577 | |
IgG |