Learn More
Cytiva (formerly Pall Lab) TF PTFE Membrane Disc Filters
Strong, chemically resistant membranes for air monitoring and sampling in aggressive environments.
Marca: Cytiva (formerly Pall Lab) 66151
Dettagli aggiuntivi : Peso : 0.31300kg
Descrizione
- Low chemical background permits highly sensitive, interference-free determinations
- Ensures accurate gravimetric determinations with low tare mass
- Ideal for filtration of gas and/or organic solvents
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
PTFE | |
0.45 μm | |
Non-sterile | |
142 mm |