Learn More
Ebro Electronic™ TFX Core Thermometer Series
Termometro preciso e molto robusto con un grande display
Marca: Ebro Electronic™ 1340-5416A
Dettagli aggiuntivi : Peso : 0.09000kg
Descrizione
Thermometer for various applications in food, laboratory, industial use. All thermometers includes a factory calibration certificate. The thermometer measures with PT 1000 temperature sensor.Specifica
EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012; EN 13485; (EC) 1935/2004 Materials and articles which come into contact with foodstuffs; The Food and Feed Hygiene and Safety Regulations 2020 2020 No. 1504; PART 3; Regulation 16 | |
Per panetterie, macellerie e industria alimentare | |
Thermometer, sensor, calibration certificate, instruction manual | |
ABS Plastic | |
Pointed | |
90 g | |
None | |
572°F | |
-58°F | |
Da –50 a 300 o da –50 a 400°C | |
AG 130, AG 140, AG 150, AG 160, AX 100 | |
Lithium 3 V/Typ CR 2477 | |
120 mm | |
22 mm |
Single-use | |
52 mm | |
54 mm | |
TFX 410-1 TPX 400 | |
0.1 °C | |
Termometro a sonda di precisione | |
With silicone cable | |
300°C | |
-50°C | |
Factory Calibrated | |
Approx. 5 Years | |
±0.3°C | |
LCD |