missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TICAM2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19524-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
TRAM, or TRIF-related adaptor molecule (TICAM2), plays an essential role in the MyD88-independent signaling of TLR4. The functional absence of TRAM in vascular endothelial cells restricts TLR4 signaling to MyD88-dependent pathway. Overexpression of TRAM cDNA in endothelial cells reconstituted LPS-induced Trif-dependent NF-kB activation and cxcl10 promoter activity.Specifica
TICAM2 | |
Polyclonal | |
Unconjugated | |
TICAM2 | |
TICAM 2, TICAM2, TIRAP3, TIRP, TRAM, TRIF related adapter molecule | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
353376 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86XR7 | |
TICAM2 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |