missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNNT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19729-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.Specifica
TNNT3 | |
Polyclonal | |
Unconjugated | |
TNNT3 | |
Fast skeletal muscle troponin T, TNNT3, Troponin T3 skeletal fast, Troponin T-FS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21957, 24838, 7140 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09739, P45378, Q9QZ47 | |
TNNT3 | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |