missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOLLIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11315-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Toll-like receptors (TLRs) are evolutionarily conserved pattern-recognition molecules resembling the toll proteins that mediate antimicrobial responses in Drosophila. These proteins recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. The TLRs act through adaptor molecules to activate various kinases and transcription factors so the organism can respond to potential infection. These adaptor molecules include TOLLIP, MyD88, and TRIF. TOLLIP associates directly with TLR2 and TLR 4, acting as an inhibitor to TLR activation. This negative regulation of TLR signaling may serve to limit the production of proinflammatory mediators during infection and inflammation.Specifica
TOLLIP | |
Polyclonal | |
Unconjugated | |
TOLLIP | |
IL 1RAcPIP, toll interacting protein, TOLLIP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361677, 54472, 54473 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2RUW1, Q9H0E2, Q9QZ06 | |
Tollip | |
TOLLIP Fusion Protein Ag1864 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |