missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Alpha-synuclein siRNA
Blocks transcription and translation of Alpha-synuclein from Human.
Marca: enQuireBio™ QX2-10nmol
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000336 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
6622 | |
10 nmol | |
Human |
SH-SY5Y. | |
SNCA | |
siRNA | |
PD1, NACP, PARK1, PARK4, SNCA | |
Lyophilized |