missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human mTOR siRNA
Blocks transcription and translation of mTOR from Human.
Marca: enQuireBio™ QX50-10nmol
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_004949 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
2475 | |
10 nmol | |
Human |
U87MG Human Glioblastoma Cells. | |
MTOR | |
siRNA | |
SKS, FRAP1, RAFT1, FRAP, RAPT1, FRAP2, Serine/threonine-protein kinase mTor, mechanistic target of rapamycin | |
Lyophilized |