missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human TLR4 siRNA
Blocks transcription and translation of TLR4 from Human.
Marca: enQuireBio™ QX30-10nmol
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_612564 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
7099 | |
10 nmol | |
Human |
HepG2 Human Carcinoma Cells from Liver. | |
TLR4 | |
siRNA | |
TLR-4, Toll-like Receptor 4, CD284 | |
Lyophilized |