missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Mouse Apoptosis regulator BAX siRNA
Blocks transcription and translation of Apoptosis regulator BAX from Mouse.
Marca: enQuireBio™ QX9-10nmol
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_031553 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
12028 | |
10 nmol | |
Mouse |
DC-1 Mouse Dendritic Cells. | |
Bax | |
siRNA | |
BCL2L4 | |
Lyophilized |