missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18807-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
TSPAN11 gene ontology annotations related to this gene include cell surface receptor signaling pathway.Specifica
TSPAN11 | |
Polyclonal | |
Unconjugated | |
TSPAN11 | |
tetraspanin 11, Tspan 11, TSPAN11, VSSW1971 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
441631, 68498 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A1L157, Q9D1D1 | |
TSPAN11 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |