Learn More
Eppendorf™ twin.tec™ PCR Plates BioBased
Scoprite le nuove piastre per PCR BioBased, realizzate con polimeri a base biologica (approccio basato sul bilancio di massa). Queste piastre consentono di ridurre in modo significativo l'impronta di carbonio legata ai materiali di consumo, pur mantenendo le stesse prestazioni tecniche.
Marca: Eppendorf™ 0030531086
Dettagli aggiuntivi : Peso : 0.87000kg
Descrizione
- Reduction of consumable-related carbon footprint in the lab
- Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
- Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
- Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
- Available with barcode (upon request)
Specifica
Custom Barcoding Available | |
LoBind | |
Colorless | |
Skirted | |
96 | |
Conical Bottom (V-well) | |
PCR Ready |
8 x 12 | |
Eppendorf | |
BioBased LoBind™ PCR Plates | |
Polypropylene, Polycarbonate | |
Non-sterile | |
25 Pk. |