Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX-90 with 2 Doors
Safe and approved storage of flammable hazardous substances in working areas
Marca: asecos™ 3070800133848
Dettagli aggiuntivi : Peso : 430.63919kg
Allerta:
For indoor use only
Descrizione
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Space saving folding doors: less required space in front of the cabinet while doors are open
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifica
1742 mm | |
Safety storage cabinet | |
450 mm | |
Powder-coated plain | |
1196 mm | |
616 x 1196 x 1968 mm | |
Left: 524 x 450 x 1742 mm, Right: 524 mmx 450 x 1742 mm | |
Tray shelf | |
1968 mm | |
S90.196.120.MV.FDAS | |
2 |
3 x shelf, 1 x perforated insert (370 mm), 1 x bottom collecting sump, 3 x tray shelf, 1 x bottom collecting sump (PP), Vertical dividing wall | |
Safe and approved storage of flammable hazardous substances in working areas | |
524 mm | |
Steel | |
Door open arrest system | |
501.50 kg | |
Light Gray | |
616 mm | |
Folding | |
6 | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |