Learn More
Cole-Parmer™ Union Threaded Elbow
Threaded fittings offer more permanent installation
Marca: Cole-Parmer™ 06546-35
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Chemically-inert PFA handles most aggressive chemicals
- Threaded reducing bushing
- Choose the size and material that meets your application needs
- Use with our rigid tubing for your high-pressure applications
Specifica
Union Threaded Elbow | |
Not Autoclavable | |
1/2 in. | |
1/2 in. NPTF x 1/2 in. NPTF |
NPT(F) Connection | |
PFA | |
Ethylene Oxide | |
Translucent |