Learn More
3M™ Universal Microfiber Wipes, Series 14
Suitable for use in hospitals, restaurants, offices, and bathrooms
Marca: 3M™ 7000061419
366.27 EUR valido fino al 2024-12-31
Usa il codice promo "21908" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.05100kg
Descrizione
- Highly versatile and reusable cloth
- Designed to simultaneously trap and remove dust, dirt, fingerprints, and moisture from a wide variety of surfaces
- Unlike conventional wipes, the ribbon-like fibers penetrate deeply into surface irregularities where dirt, grease, and micro-organisms hide
- Machine washable up to 90°C
- Resists up to 300 wash cycles
Specifica
Wipes | |
400 mm | |
Blue | |
Cleaning Common Surfaces | |
Machine Washable |
400 mm | |
Microfiber | |
400 x 400 mm | |
10/Pk. |