Learn More
Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers
Ensure compliance with regulatory guidelines with the Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers providing tools and documentation for system qualification.
Marca: Thermo Scientific™ 840-293000
Dettagli aggiuntivi : Peso : 1.20000kg
Descrizione
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ) with Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers. Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer Validation Solutions include:- Information on the validation process
- Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
- Information on pharmacopoeia recommended tests for specific instrument performance where applicable
- Installation and instrument certification
Solutions available for GENESYS 40-180, BioMate 160, Evolution200 series, Evolution 350, NanoDrop One, NanoDrop 2000, and NanoDrop 8000 instruments
Specifica
Validator 2 for INSIGHT – A4 Version | |
Evolution 200 series and Evolution 350 Spectrophotometers |