Learn More
Ansell™ Vantage™ 70-750 Series Yellow and Blue Aramid Fiber Heavy Weight Gloves
Optimal combination of comfort and dexterity, maximum cut protection combined with superior longevity.
Marca: Ansell™ 70750080
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Heavy duty gloves
- Use innovative technology for improved cut resistance, excellent flexibility
- Higher level of user comfort
- Revolutionary engineered yarns with the highest cut-protection-to-weight ratio
- Ideal for handling sharp tools and materials
- Includes protection from heat and light molten splashes
Applications:
- Assembling metal parts and components
- Body assembly
- Press and stamping
- Handling sharp or irregular-edged metal parts
- Protection from heat and light molten splashes
- Welding
- Default checking
- Metal finish inspection
- Moderately oily metal parts with sharp or rough edges
- Automated welding feeding
Specifica
Dupont™ Kevlar™ aramid fibers | |
Dupont™ Kevlar™ aramid fibers | |
8 | |
Dupont™ Kevlar™ aramid fibers | |
12 Pairs |
CE Mark, EN 388 : 154x, EN 407 | |
Heavyweight Gloves | |
Knit | |
Dupont™ Kevlar™ aramid fibers | |
High cut resistance |