Learn More
Emma Safety Footwear Vaya Safety Shoes
Easy to put on, allowing you to change shoes without bending over. The upper is fully washable unique microfiber. Features SmartecAir™ for dry, fresh feet while the protective toe cap prevents staining and is lubricant- and fat-resistant.
Marca: Emma Safety Footwear MM50054444
Dettagli aggiuntivi : Peso : 1.54100kg
Descrizione
- S2-Safety shoe for wet working conditions
- Microfiber construction
- GripForce™ Easy Twist
- Chrome-free lining
- Heat and cold insulated
- Sanitized
Specifica
EN ISO 20345:2011 | |
White | |
Hydro-Tec™ Sanitized Silver | |
44 |
Elastic | |
Vaya | |
Microfiber | |
Steel |