Learn More
Sartorius Stedim Biotech™ Venema Rolls, Qualitative and Technical Papers, Smooth, Grade 100/N
Sartorius Stedim Biotech™ Venema Rolls, Qualitative and Technical Papers, Smooth, Grade 100/N are suitable for analytical and technical purposes and commonly used in the sugar industry.
Marca: Sartorius Stedim Biotech™ FT-1-328-2401000
Dettagli aggiuntivi : Peso : 21.20000kg
Descrizione
- Suitable for use in the sugar industry
- Suitable for use with Venema systems
- Wet strengthened and smooth
- Cellulose construction
Specifica
85 g/m2 | |
Assaying sugar | |
1 Roll | |
1000m x 240 mm | |
240 mm |
Venema roll | |
100/N | |
0.18 mm | |
1000 m |