Learn More
Saint Gobain Life Sciences Tappo silicone Versilic™
Use these stoppers for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Versilic™ Silicone Caps are particularly suited for all types of tissue culture work.
Marca: Saint Gobain Life Sciences 261014
501.73 EUR valido fino al 2024-04-30
Usa il codice promo "21637" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Biologically neutral and non-toxic silicone caps for closing tubes and flasks
- Odorless, tasteless, non-adherent and resistant to aging
- Nontoxic, biologically neutral
- FDA/BGA approved
- For test tubes
- Easy to install
Specifica
Silicone Stopper | |
14 mm | |
Silicone | |
Autoclavable |
6 mm | |
500 Pk. | |
-70 to +250°C | |
FDA Compliant Food Contact, BFR, BGA |