Learn More
VistaLab™ Technologies Nozzle Filter Replacement Kits
Nozzle filter kit for Macro model 0.2-10μL
Marca:  VistaLab™ Technologies 9060-5009
Dettagli aggiuntivi : Peso : 0.00100kg
Codice prodotto. 11916052
  • 19.00€ / pezzi 10
Spedizione stimata: 19-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Replacement nozzle contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into the pipet
  • If filter becomes wet, pipet will not aspirate fluid until a new filter or replacement nozzle is installed
Specifica
Specifica
Non-sterile
100 to 5000 μL Macro Pipette
5 in.
1 in.
5 mL
10/Bag
3 in.