Learn More
VistaLab™ Technologies Nozzle Filter Replacement Kits
Nozzle filter kit for Macro model 0.2-10μL
Marca: VistaLab™ Technologies 9060-5009
Dettagli aggiuntivi : Peso : 0.00100kg
Descrizione
- Replacement nozzle contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into the pipet
- If filter becomes wet, pipet will not aspirate fluid until a new filter or replacement nozzle is installed
Specifica
Non-sterile | |
100 to 5000 μL Macro Pipette | |
5 in. | |
1 in. |
5 mL | |
10/Bag | |
3 in. |