Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Single Certificate
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Single Certificate is ideal for measuring and transferring various quantities of liquid.
Marca: Poulten & Graf GmbH R385/WAC/C/5
Dettagli aggiuntivi : Peso : 0.07500kg
Descrizione
- Total delivery
- Conformity approved
- Zero at top
- Amber graduation
- With ring marks
- Non blowout
- With single certificate
Specifica
Volac Fortuna Graduated pipette, one mark | |
EN ISO 835 | |
No | |
0.03mL | |
Yes |
Soda Lime Glass | |
AS | |
5 mL | |
0.05 mL |