missing translation for 'onlineSavingsMsg'
Learn More

YARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17150-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16896984

  • 121.38€ / 20µL

Spedizione stimata: 05-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes a mitochondrial protein that catalyzes the attachment of tyrosine to tRNA(Tyr). Mutations in this gene are associated with myopathy with lactic acidosis and sideroblastic anemia type 2 (MLASA2).
Specifica

Specifica

YARS2
Polyclonal
Unconjugated
YARS2
CGI 04, mt TyrRS, Tyrosine tRNA ligase, TYRRS, YARS2
Rabbit
Antigen Affinity Chromatography
RUO
287924, 51067, 70120
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5I0L3, Q8BYL4, Q9Y2Z4
Yars2
YARS2 Fusion Protein Ag9530
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali