Learn More
MP Biomedicals™ Z-VAD(OMe)-Fluromethylketone Inhibitor
The Z-VAD(OMe)-FMK inhibitor synthesized using proprietary technology is cell permeable and irreversibly binds to activated Caspases to block apoptosis.
Brand: MP Biomedicals™ 03FK00903
Additional Details : Peso : 0.01000kg
Description
Z-VAD(OMe)-FMK is an irreversible and cell permeable broad-spectrum Caspase Inhibitor. It has been shown to inhibit Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine).
Specifications
White | |
3 mg | |
467.487 |
4°C | |
Z-Val-Ala-Asp-(OMe)-Fluromethylketone Inhibitor | |
Solid |