Learn More
Emma Safety Footwear Zandvoort Safety Shoes
A mid-level boot with water repellant oiled Nubuck uppers, internal steel toe protection and an exceptional sticky grip even on wet and greasy surfaces. Primarily designed for Metal workers, the Zandvoort is used in outdoor occupations where a strong, safe, comfortable shoe is essential.
Marca: Emma Safety Footwear MM53884648
Dettagli aggiuntivi : Peso : 2.20500kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- Oil Nubuck construction
- GripForce™ Duo Heat Resistant Outsole
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifica
EN ISO 20345:2011 | |
Zandvoort | |
Oiled Nubuck |
Brown | |
Full grain Nappa leather / Split leather | |
48 |