missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF747 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19901-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
ZNF747 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.Specifica
ZNF747 | |
Polyclonal | |
Unconjugated | |
ZNF747 | |
zinc finger protein 747, ZNF747 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
65988 | |
-20°C | |
Liquid |
Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BV97 | |
ZNF747 | |
ZNF747 Fusion Protein Ag13821 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |