New Feature
You can now browse the site in English or Español.
DNA Extraction Purification Product Image
DNA Extraction and Purification
The isolation of DNA from different sample types and its subsequent purification requires a range of reagents and consumables. We offer a comprehensive portfolio of kits and chemicals for plasmid (miniprep, midiprep, maxiprep), PCR, and genomic DNA purification, as well as gel extraction and more.
FEATURED
MP Biomedicals FastDNA™ SPIN Kit for Soil
Quickly and efficiently isolates bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples.
DNA Extraction Purification Product Image 2
Invitrogen™ PureLink™ Microbiome DNA Purification Kit
Enables fast purification of high-quality microbial and host DNA
A microbiome DNA purification kit with proven PureLink™ spin column technology for robust yields of purified DNA.
DNA Extraction Purification Product Image 3
Greener Choice DNA Extraction and Purification
Reduce your environmental impact with our selection of Greener Choice products
Kits, reagents, and other supplies for the extraction of DNA from a sample and its subsequent purification.
DNA Extraction Purification Product Image 4
Save Now - Exclusive Deals
Complete Your Order - Great Deals
FAQ
The purpose of DNA extraction and purification is to isolate DNA from cells or tissues so that it can be used for various downstream applications. These applications include:
  • Genetic Research: Studying the structure, function, and regulation of genes
  • Diagnostics: Identifying genetic disorders, infectious diseases, and other health conditions
  • Forensic Analysis: Analyzing DNA from crime scenes for identification purposes
  • Biotechnology: Genetic engineering, cloning, and production of genetically modified organisms (GMOs)
  • Pharmaceutical Development: Discovering and developing new drugs and therapies
  • Agricultural Science: Breeding plants and animals with desirable traits
DNA extraction and purification ensure that the DNA obtained is free from contaminants such as proteins, lipids, and other cellular components that could interfere with these applications.
To purify DNA after extraction, common methods include:
  1. Phenol-Chloroform Extraction: Removes proteins using phenol and chloroform, followed by ethanol precipitation.
  2. Ethanol Precipitation: DNA is precipitated with ethanol and sodium acetate, then washed with 70% ethanol.
  3. Silica Column-Based Methods: DNA binds to silica columns, contaminants are washed away, and DNA is eluted.
  4. Magnetic Bead-Based Methods: DNA binds to magnetic beads, contaminants are washed away, and DNA is eluted.
  5. Cesium Chloride Density Gradient Centrifugation: Uses ultracentrifugation to separate DNA based on density.
Each method is chosen based on the purity requirements and resources available.
Filtering is important in DNA extraction for several reasons:
  • Removal of Debris: It helps to eliminate particulate matter, such as cell fragments and undissolved materials, that could interfere with subsequent steps in the extraction process
  • Improving Purity: By removing contaminants, filtering ensures that the extracted DNA is cleaner and more suitable for downstream applications like PCR, sequencing, and cloning
  • Preventing Equipment Damage: Filtering protects laboratory equipment (e.g., centrifuges, pipettes) from clogging and potential damage caused by debris
  • Enhancing Accuracy: Cleaner samples lead to more accurate and reliable results in genetic analysis and other applications
Overall, filtering is a crucial step to ensure the quality and integrity of the extracted DNA.
Resources and Related Information
Improve Microbiome Studies
Select products for your microbiome research.
25355 Merch P8 Doc 1
FastPrep® System A Complete Solution for Your Sample Preparation
A detailed guide to FastPrep instruments, tube holders, Lysing Matrix, Nucleic Acid and Protein Purification kits, and other tools for achieving high yields of DNA and RNA.
Labels and Ribbons
Spotlight on Magnetic Beads
Magnetic beads - versatile super (paramagnetic) particles well suited to DNA extraction, size selection, and clean-up as alternatives to column-based purification.
Labels and Ribbons
Shop By Brand