Learn More
Dräger™ CMS Chips: Benzene
Suitable for 10 measurements of one gas
Marca: Dräger™ 6406030
Dettagli aggiuntivi : Peso : 0.02700kg
Descrizione
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.Specifica
700 to 1100 hpa | |
1 to 30mg/L | |
35 to 300 sec. | |
0°C to 40°C |
Benzene CMS Chip | |
0.2 to 10 | |
32°F to 104°F |