Learn More
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps is a gas collecting tube with O-ring valves on each end.
Marca: GPE Scientific™ GST/2000
Dettagli aggiuntivi : Peso : 0.40000kg
Descrizione
- Cylindrical, oblong shape
- Gas tight container
- Gauged volume
- Standard collecting tube with valved ends
Specifica
Gas Sample Tube | |
2000 mL |