Circulating Baths
Circulating baths are constant temperature water baths that enable rapid heating and cooling of samples by constantly circulating water. They are often used for laboratory or industrial applications where a wide range of temperature control is needed.
Fisherbrand™ Isotemp™ R20 Refrigerated and Heated Bath Circulators
Low-profile model enables easy bath access when lab counter height presents a challenge. Seamless stainless-steel tanks guarantee durability and easy cleaning.
Contec™ Sterile IPA
Fisherbrand™ Isotemp™ Heated Bath Circulators: Model 4100 H7
Reliable, high-wattage heaters that deliver rapid fluid heat up and ensure fluid compatibility for your application. Standard high temperature cutout (HTC) circuitry is user-selectable to ensure application temperatures do not exceed set point parameters.
Thermo Scientific™ ARCTIC A10 Refrigerated Circulators
A10 refrigerated bath circulators provide efficient cooling for open and closed applications, with temperatures ranging from -10° to +100°C and 6L maximum bath volume.
FAQ
A circulating bath maintains a stable set temperature for samples and instruments by moving fluid through a bath (and often an external loop). It’s commonly used for reaction control, viscosity testing, calibration, and temperature conditioning. Options from Julabo, Thermo Scientific, Huber, Fisherbrand, and IKA support different ranges and workloads.
Choose heating-only if you only need temperatures above ambient. Pick a refrigerated circulator if you must control below ambient or offset heat from connected equipment. Compare temperature range, cooling capacity, and pump performance. Brands like Julabo, Thermo Scientific, Huber, Fisherbrand, and IKA offer both classes for typical lab demands.
Temperature stability depends on control accuracy, bath fluid circulation, and thermal load. Look for strong pump flow/pressure, suitable bath volume, and adequate heating/cooling power for your setpoint. Also consider insulation and lid options to reduce losses. Julabo, Thermo Scientific, Huber, Fisherbrand, and IKA models vary by performance tier.
Use the circulator’s inlet/outlet ports with compatible tubing rated for your temperature range and pressure. Prime the loop, remove air, and ensure the pump setting matches the restriction of your system. Many Julabo, Thermo Scientific, Huber, Fisherbrand, and IKA units support external circulation for jackets, condensers, and instruments.
Top Brands