Speciality Pipette Tips
Speciality pipette tips are disposable and for specific applications and tasks such as transferring liquids between vials. Browse through a broad range from leading brands. Choose from filtered, non-filtered, positive displacement, stepper, multi-stepper and repeater tips depending on your need.
FEATURED
Gilson™ MICROMAN™ E Capillary Pistons
MICROMAN™ E Capillary Pistons provides reproducible, reliable results even with the most viscous or volatile liquids.
Contec™ Sterile IPA
EASY-TO-READ
Gilson™ MICROMAN™ Capillary Pistons
Precision handling for viscous liquids.
Contec™ NeutraKlean™ Detergent with Purified Water, Sterile
READY-TO-USE
Eppendorf™ Combitips™ Advanced Pipettor Tips
Versatile tips for reliable pipetting.
Contec™ Non-Sterile CyChlor
Complete Your Order - Great Deals
Multipette™ E3 Single Channel Pipet
Dispenses and pipets any liquid regardless of viscosity while maintaining the highest standards for accurate results. Eppendorf™ Multipette™ E3 Single Channel Pipet provides full volume control, improved accuracy, and reproducibility while reducing physical strain during use.
Universal Pipette Tips
Fisherbrand™ Polypropylene Dispenser Tips: Adapter
Polypropylene made adapter for dispenser tip 25/50 mL that is ideal in most research, scientific, and laboratory environments.
Pipette Specific Tips
Some Tips on Tips
The performance of a pipette is related to its tip and we can help you find one that fits perfectly.
Pipette Specific Tips
FAQ
Specialty pipette tips offer several benefits in a laboratory setting, including enhanced precision and accuracy for specific applications. They are designed to meet the unique requirements of various experiments, such as low-retention tips for viscous liquids or filter tips for contamination prevention. Specialty tips can improve the efficiency of workflows by providing better fit and compatibility with specific pipettes, reducing the risk of leaks and ensuring consistent sample dispensing. Additionally, they can help minimize sample loss and cross-contamination, which is crucial for sensitive assays and experiments.
Proper maintenance and storage of specialty pipette tips is essential for ensuring their performance and longevity. Store tips in a clean, dry environment to prevent contamination and damage. Keep them in their original packaging until use to maintain sterility and prevent exposure to dust and other contaminants. Regularly inspect tips for any signs of damage or defects before use. Avoid touching the tips with bare hands to prevent contamination. If using filter tips, ensure that the filters are intact and not compromised. Proper handling and storage practices will help maintain the integrity and reliability of specialty pipette tips.
Common issues with specialty pipette tips include improper fit or compatibility with pipettes, which can lead to leaks or inconsistent volume dispensing. Ensure that the tips are compatible with the pipette model being used and are properly attached. Contamination is another issue, especially with filter tips; always handle tips with care and store them properly to maintain sterility. Clogging can occur with certain types of tips, such as filter tips, if not used correctly. Regularly inspect and replace tips as needed to prevent clogging. By addressing these common issues through proper selection, handling, and storage, you can ensure the reliable performance of your specialty pipette tips.
Shop By Brand