Microcentrifuges
Microcentrifuges are essential in molecular biology, clinical, pharmaceutical and industrial laboratories for rapid separation of small-volume samples. From quick spin-downs of nucleic acids and proteins to plate-based assays and haematocrit determinations, choosing the right rotor format, speed and footprint ensures reliable results and efficient workflows.
Make Us an Offer on Centrifuges
Unlock special pricing on top centrifuges. Make us an offer today and upgrade your lab equipment. Limited time only!
Contec™ Sterile IPA
Thermo Scientific™ mySPIN™ 6 Mini Centrifuge
Our most compact centrifuge, the mySPIN 6 mini centrifuge, is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microcentrifuge tubes.
Contec™ Sterile IPA
OHAUS™ Frontier™ 5513 Series Micro-Centrifuges
Micro centrifuges have five optional rotors designed for clinical, industrial, and research applications.
Contec™ Sterile IPA
Save Now - Exclusive Deals
FAQ
A microcentrifuge is a compact benchtop centrifuge designed for small-volume tubes or strips, typically up to 2.0 mL. It is used for pelleting nucleic acids, proteins, cells and reaction components in PCR, cloning and clinical workflows using instruments from brands such as Thermo Scientific, Fisherbrand, Corning and Ohaus.
Mini microcentrifuges are ideal for quick spin-downs and routine tasks at ambient temperature, while refrigerated models maintain set temperatures for sensitive enzymes, nucleic acids and clinical samples. Choose based on run time, target temperature, rotor capacity and whether your protocols specify cooling, for example with Sorvall or Axygen instruments.
Common rotor types include fixed-angle rotors for 1.5/2.0 mL tubes, strip rotors for PCR strips, swing-out rotors for microplates and dedicated hematocrit rotors. Many Thermo Scientific, Fisherbrand and Ohaus systems offer interchangeable rotors, allowing you to expand from basic pelleting to plate-based assays without changing instruments.
G-force (RCF) and RPM determine how quickly a centrifuge pellets particles at a given radius. Instruments such as the Ohaus FC5513 or Sorvall Legend Micro series provide high RCF values for fast separations, while mini models focus on convenience. Matching protocol-specified g-force helps ensure reproducible yields and consistent separations across runs.
Always balance tubes by volume and weight, close biocontainment lids securely and select rotors rated for your maximum RPM and g-force. Using sealed rotors, such as ClickSeal systems on Fisherbrand and Thermo Scientific models, helps minimise aerosol risks, while regular maintenance and cleaning preserve performance and extend instrument lifetime.
Resources and Related Information
Eppendorf™ QuickLock Technology in Action
Top Brands