Round Plastic Bottles
Round plastic bottles are a core choice for liquid storage, sample collection and transport across chemistry, life science and environmental labs. This range from Thermo Scientific Nalgene, Fisherbrand, Azlon, Kautex and Gosselin covers volumes from small sample vials to multi-litre containers in HDPE and LDPE materials. Options include wide-neck designs for easy filling, leak-resistant closures, graduated walls and robust construction for demanding workflows. Use these bottles for buffer and media storage, reagent preparation, waste collection and shipping of water or soil samples while supporting consistent, reproducible results across sites and teams.
Useful Links
Save Now - Exclusive Deals
Product Code 10623141
Product Code 10515733
Product Code 10074810
Product Code 10664212
Product Code 10569831
Product Code 15241609
Product Code 10505021
Product Code 10414501
Product Code 10102881
Product Code 10411601
Product Code 12632415
Product Code 10116230
Product Code 10633141
Complete Your Order - Great Deals
Product Code 10196780
Product Code 10657681
Product Code 10404501
Must Have
Product Code 11962659
Product Code 11713094
Product Code 10477043
Product Code 10176590
Product Code 10360025
Product Code 10167120
Product Code 10353501
Product Code 10652751
Product Code 11769813
Product Code 11324455
Product Code 10058240
Product Code 10508272
FAQ
Most round plastic bottles in this range are manufactured from HDPE or LDPE. HDPE bottles from Thermo Scientific Nalgene, Fisherbrand and Azlon provide good chemical resistance and rigidity, while LDPE bottles from Kautex and Gosselin offer improved flexibility and reliable squeeze control.
Select bottle volume according to the working volume you need plus headspace for expansion, mixing or freezing. Smaller Fisherbrand and Gosselin bottles help minimise reagent waste, while larger Thermo Scientific Nalgene and Azlon containers suit bulk buffer storage or transport of environmental samples.
Round plastic bottles are available with standard screw caps, wide-mouth leak-resistant caps and specialised pouring or dispensing closures. Thermo Scientific Nalgene, Fisherbrand, Azlon, Kautex and Gosselin offer compatible caps designed to maintain a secure, reliable seal during storage and transport.
Check the chemical compatibility of HDPE or LDPE with your solvent or reagent using manufacturer data from Thermo Scientific Nalgene, Azlon or Kautex. For aggressive chemicals, verify maximum concentration, temperature limits and contact times to avoid swelling, cracking or permeation of the bottle walls.
Branded round plastic bottles from Thermo Scientific Nalgene, Fisherbrand, Azlon, Kautex and Gosselin typically offer better batch consistency, traceability and validated performance. This supports standardised storage procedures, reproducible results and a lower risk of leaks or contamination during transport and handling.